Learn More
Abcam SC 221409, Cdk2 inhibitor, 5MG
Catalog No. 502705442
Encompass_Preferred
MW 395.23 g/mol, Purity >97%. Potent, selective, ATP-competitive Cdk2 inhibitor (IC₅₀ = 60 nM). Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5442 Supplier Abcam Supplier No. AB1450535MG
Add to Cart
Add to Cart
MW 395.23 g/mol, Purity >97%. Potent, selective, ATP-competitive Cdk2 inhibitor (IC₅₀ = 60 nM). Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement