Learn More
Medchemexpress LLC HY-W010983 5mg Medchemexpress, SC-236 CAS:170569-86-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0109835MG
Encompass_Preferred
Medchemexpress, HY-W010983 5mg SC-236 CAS:170569-86-5 SC-236 is an orally active COX-2 specific inhibitor (IC50 = 10 nM) and a PPARγ agonist. SC-236 suppresses activator protein-1 (AP-1) through c-Jun NH2-terminal kinase. SC-236 exerts anti-inflammatory effects by suppressing phosphorylation of ERK in a murine model[4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3305