Learn More
MVE BIOLOGICAL SOLUTIONS MVE SC 4/3V CryoShipper, QWick Charge Technology, Lightweight Aluminum Construction, 4.5 L LN2 Capacity
Catalog No. NC1929464
Encompass_Preferred
The MVE SC 4/3V CryoShipper is designed for the safe transport of biological samples at cryogenic temperatures of -150°C or colder. Its QWick Charge Technology allows for quick liquid nitrogen charging for efficient vapor shipping. Constructed from lightweight aluminum with advanced protection technologies, it ensures reliability and sample security during transit, making it a leading choice for cryogenic shipping needs.
Catalog No. NC1929464 Supplier MVE BIOLOGICAL SOLUTIONS Supplier No. 10817330
The MVE SC 4/3V CryoShipper is designed for the safe transport of biological samples at cryogenic temperatures of -150°C or colder. Its QWick Charge Technology allows for quick liquid nitrogen charging for efficient vapor shipping. Constructed from lightweight aluminum with advanced protection technologies, it ensures reliability and sample security during transit, making it a leading choice for cryogenic shipping needs.