Learn More
Enzo Life Sciences SC-51322 (25 mg). CAS: 146032-79-3
SDP
Catalog No. 502011717
Encompass_Preferred
A potent and selective prostaglandin E2 antagonist (pA2=8.1). Displays in vivo analgesic activity in the mouse (ED50=0.9 mg/kg). Purity: ≥98% (HPLC). Solubility: Soluble in ethanol (10mg/ml) or DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1717 Supplier Enzo Life Sciences Supplier No. BMLRA1120025
A potent and selective prostaglandin E2 antagonist (pA2=8.1). Displays in vivo analgesic activity in the mouse (ED50=0.9 mg/kg). Purity: ≥98% (HPLC). Solubility: Soluble in ethanol (10mg/ml) or DMSO (25mg/ml). Long Term Storage: -20°C.