Learn More
Enzo Life Sciences SC-514 (10mg). CAS: 354812-17-2
SDP
Supplier:  Enzo Life Sciences BMLEI3430010
Encompass_Preferred
Selective, reversible and ATP-competitive inhibitor of IKK2 (IC50=11.2µM). Does not inhibit other IKK isoforms or other serine/threonine or tyrosine kinases. Inhibits transcription of NF-κB dependent genes in IL-1β-induced rheumatoid arthritis-derived synovial fibroblasts in a dose-dependent fashion. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0728