Learn More
Medchemexpress LLC HY-13802 50mg , SC-514 GK 01140 CAS:354812-17-2 Purity:98%
SDP
Catalog No. 501493946
Encompass_Preferred
Medchemexpress, HY-13802 50mg SC-514 GK 01140 CAS:354812-17-2 Formula:C9H8N2OS2 IC50: 11.2±4.7 μM (IKK-2), 2.7±0.7 μM (Recombinant human IKK-1/IKK-2 heterodimer), 6.1±2.2 μM (Native IKK complex), 61 μM (CDK2/A), 71 μM (AUR2), 75 μM (PRAK), 123 μM (MSK), >200 μM (IKK-1), >200 μM (TBK-1) Purity:98% SC-514 is a selective IKK-2 inhibitor ( IC 50 =11.2±4.7 μM), which does not inhibit other IKK isoforms or other serine-threonine and tyrosine kinases. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3946 Supplier Medchemexpress LLC Supplier No. HY1380250MG
Medchemexpress, HY-13802 50mg SC-514 GK 01140 CAS:354812-17-2 Formula:C9H8N2OS2 IC50: 11.2±4.7 μM (IKK-2), 2.7±0.7 μM (Recombinant human IKK-1/IKK-2 heterodimer), 6.1±2.2 μM (Native IKK complex), 61 μM (CDK2/A), 71 μM (AUR2), 75 μM (PRAK), 123 μM (MSK), >200 μM (IKK-1), >200 μM (TBK-1) Purity:98% SC-514 is a selective IKK-2 inhibitor ( IC 50 =11.2±4.7 μM), which does not inhibit other IKK isoforms or other serine-threonine and tyrosine kinases. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.