Learn More
Abcam SC-514, IKKbeta inhibitor, 25MG
Catalog No. 502705772
Encompass_Preferred
MW 224.3 Da, Purity >98%. Selective, ATP-competitive, irreversible IKKβ inhibitor (IC₅₀ = 3-12 μM). Reduces iNOS induction. Shows anti-inflammatory effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5772 Supplier Abcam Supplier No. AB14441525MG
MW 224.3 Da, Purity >98%. Selective, ATP-competitive, irreversible IKKβ inhibitor (IC₅₀ = 3-12 μM). Reduces iNOS induction. Shows anti-inflammatory effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement