Learn More
Abcam SC-514, IKKbeta inhibitor, 5MG
Catalog No. 502705770
Encompass_Preferred
MW 224.3 Da, Purity >98%. Selective, ATP-competitive, irreversible IKKβ inhibitor (IC₅₀ = 3-12 μM). Reduces iNOS induction. Shows anti-inflammatory effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5770 Supplier Abcam Supplier No. AB1444155MG
MW 224.3 Da, Purity >98%. Selective, ATP-competitive, irreversible IKKβ inhibitor (IC₅₀ = 3-12 μM). Reduces iNOS induction. Shows anti-inflammatory effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement