Learn More
Medchemexpress LLC HY-59105 5mg Medchemexpress, SC-560 CAS:188817-13-2 Purity:>98%
SDP
Catalog No. 502082263
Encompass_Preferred
Medchemexpress, HY-59105 5mg SC-560 CAS:188817-13-2 SC-560 is a potent and selective COX-1 inhibitor with an IC50 of 9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2263 Supplier Medchemexpress LLC Supplier No. HY591055MG
Medchemexpress, HY-59105 5mg SC-560 CAS:188817-13-2 SC-560 is a potent and selective COX-1 inhibitor with an IC50 of 9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.