Learn More
Perkin Elmer US LLC SC Autosampler Probe PTFE 1.0 mm I.D.
Catalog No. NC0551459
Encompass_Preferred
SC Autosampler Probe, Carbon fiber support, 1.0mm ID (gray marker), long probe, long capillary, connects to port #5 (1/4-28 fitting) high flow.
Catalog No. NC0551459 Supplier Perkin Elmer US LLC Supplier No. N0777522
SC Autosampler Probe, Carbon fiber support, 1.0mm ID (gray marker), long probe, long capillary, connects to port #5 (1/4-28 fitting) high flow.