Learn More
Perkin Elmer US LLC SC Autosampler Probe PTFE 0.8 mm I.D.
Catalog No. 502105268
Encompass_Preferred
Probe with attached sample tubing, 2 probe clamps, 2 thumbscrews, 5 O-rings Probe Length: 10 inches (25.4 cm) Capillary Length: 108 (2.74 m) Probe Material: FEP-covered carbon fiber Capillary Material: PFA
Catalog No. 50-210-5268 Supplier Perkin Elmer US LLC Supplier No. N0777523
Probe with attached sample tubing, 2 probe clamps, 2 thumbscrews, 5 O-rings Probe Length: 10 inches (25.4 cm) Capillary Length: 108 (2.74 m) Probe Material: FEP-covered carbon fiber Capillary Material: PFA