Learn More
MSC SC Johnson Professional Hand Sanitizer, Foam, 80% Ethyl Alcohol, 400 mL
Catalog No. 5000508767
Encompass
This highly effective, perfume-free, and dye-free alcohol-based foam hand sanitizer kills up to 99.9999% of common germs in 15 seconds. Its innovative foam formula ensures complete coverage without mess, ideal for critical hand hygiene environments like cruise ships, food manufacturing, and long-term care.
  • Kills up to 99.9999% of many common germs in 15 seconds.
  • Foaming action prevents dripping and splashing, ensuring complete control.
  • Contains no gelling agents, leaving no sticky residue.
  • Pleasant for frequent use, encouraging compliance.
  • Non-drying formula maintains skin moisture.
  • Glove compatible for use prior to wearing gloves.
  • Dermatologist tested, hypoallergenic for sensitive skin.
  • Perfume-free and dye-free to reduce dermal sensitization.
  • Suitable for food handling; does not affect food quality or safety.
Catalog No. 50-005-08767 Supplier MSC Supplier No. 31579121
This highly effective, perfume-free, and dye-free alcohol-based foam hand sanitizer kills up to 99.9999% of common germs in 15 seconds. Its innovative foam formula ensures complete coverage without mess, ideal for critical hand hygiene environments like cruise ships, food manufacturing, and long-term care.
  • Kills up to 99.9999% of many common germs in 15 seconds.
  • Foaming action prevents dripping and splashing, ensuring complete control.
  • Contains no gelling agents, leaving no sticky residue.
  • Pleasant for frequent use, encouraging compliance.
  • Non-drying formula maintains skin moisture.
  • Glove compatible for use prior to wearing gloves.
  • Dermatologist tested, hypoallergenic for sensitive skin.
  • Perfume-free and dye-free to reduce dermal sensitization.
  • Suitable for food handling; does not affect food quality or safety.