Learn More
Sarstedt Inc SC MTUBE 2ML MDC ASSAY/PK1000
Catalog No. NC9044906 Shop All Sarstedt Inc. Products
Encompass_Preferred
Microtube Screw Cap 2ml Conical Non-Skirted Bottom for Use with Molecular Devices Threshold Kit PCR Performance Certified
Catalog No. NC9044906 Supplier Sarstedt Inc Supplier No. 72.693.465
Microtube Screw Cap 2ml Conical Non-Skirted Bottom for Use with Molecular Devices Threshold Kit PCR Performance Certified