Learn More
Sunrise Science Products SC Powder, 30 g
Catalog No. NC0772222
Encompass
Commonly added to YNB, nitrogen and glucose, at suggested g/L, for a complete yeast media.
Catalog No. NC0772222 Supplier Sunrise Science Products Supplier No. 1300030
Commonly added to YNB, nitrogen and glucose, at suggested g/L, for a complete yeast media.