Learn More
Sarstedt Inc SC TUBE CBS 20X76SPOONST/PK500
Supplier:  Sarstedt Inc 80.734.001
Encompass_Preferred
Screw Cap Feces Tube Conical Bottom Skirted 20x76mm Polypropylene Cap with Spoon Sterile
Catalog No. NC9574115