Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SC-Ura (Powder)

Catalog No. MP114410632 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP114410632 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114410632 Supplier MP Biomedicals™ Supplier No. 114410632
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Also known as Hopkins Mixture. MP Biomedicals SC-Ura (Powder) is a single dropout formulation of Synthetic Complete Supplement Mixture of amino acids for S. cerevisiae growth media. SC-Ura medium creates a richer medium for better yield and increases the

The selection of plasmids in yeast is based on the use of auxotrophic mutant strains that cannot grow without a specific media component (an amino acid, purine or pyrimidine). Transformation with a plasmid containing the mutated gene enables the transformant to grow on a medium lacking the required component.

SC-Ura medium create a richer medium for better yield and growth rate, and increase the probability of successful transformations when screening libraries or performing gene knock-outs. To use, add 1.9g of SC-Ura to each liter of DOB or DOBA and autoclave at 121°C for 15 minutes for complete synthetic defined medium for S.cerevisiae.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.