Learn More
Sunrise Science Products SC-Ura Powder, 30 g
Supplier:  Sunrise Science Products 1306030
Encompass
Commonly added to YNB, nitrogen and glucose, at suggested g/L, for a complete yeast media.
Catalog No. NC0893749