Learn More
Hach Company SC1000 4 Sens 4 mA Profibus DP
Catalog No. 5000207395 Shop All Hach Company Products
Encompass
SC1000 Probe Module-4 Sensor Connectors-4 mA Output-4 mA/digital Input-Profibus DP-4 Relays-100-240 VAC with ConduitsIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual
Catalog No. 50-002-07395 Supplier Hach Company Supplier No. LXV400.99.1F572
SC1000 Probe Module-4 Sensor Connectors-4 mA Output-4 mA/digital Input-Profibus DP-4 Relays-100-240 VAC with ConduitsIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual