Learn More
Hach Company SC1000 4 Sens 8mA Out Modbus
Catalog No. 5000207396 Shop All Hach Company Products
Encompass
SC1000 Probe Module-4 Sensor Connectors-8 mA Output-Modbus 485-100-240 VAC-with US plugIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual-US Power Cord
Catalog No. 50-002-07396 Supplier Hach Company Supplier No. LXV400.99.5A322
SC1000 Probe Module-4 Sensor Connectors-8 mA Output-Modbus 485-100-240 VAC-with US plugIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual-US Power Cord