Learn More
Hach Company SC1000 4 Sensor 4mA Out Modbus
Catalog No. 5000207943 Shop All Hach Company Products
Encompass
SC1000 Probe Module-4 Sensor Connectors-4 mA Output-4 mA/digital Input-Modbus 485-4 Relays-100-240 VAC-with US plugIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual-US Power Cord
Catalog No. 50-002-07943 Supplier Hach Company Supplier No. LXV400.99.5B522
SC1000 Probe Module-4 Sensor Connectors-4 mA Output-4 mA/digital Input-Modbus 485-4 Relays-100-240 VAC-with US plugIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual-US Power Cord