Learn More
Hach Company SC1000 6 Sens Progno Modbus485
Catalog No. 5000207905 Shop All Hach Company Products
Encompass
SC1000 Probe Module-6 Sensor Connectors-Prognosys-Modbus 485-100-240 VAC with ConduitsIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual
Catalog No. 50-002-07905 Supplier Hach Company Supplier No. LXV400.99.1H082
SC1000 Probe Module-6 Sensor Connectors-Prognosys-Modbus 485-100-240 VAC with ConduitsIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual