Learn More
Hach Company SC1000 6 Sensor 4mA Out 4mA In
Catalog No. 5000208030 Shop All Hach Company Products
Encompass
SC1000 Probe Module-6 Sensor Connectors-4 mA Output-4 mA/digital Input-4 Relays-100-240 VAC with ConduitsIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual
Catalog No. 50-002-08030 Supplier Hach Company Supplier No. LXV400.99.1R582
SC1000 Probe Module-6 Sensor Connectors-4 mA Output-4 mA/digital Input-4 Relays-100-240 VAC with ConduitsIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual