Learn More
Hach Company SC1000 6 Sensor 8mA Modbus 485
Catalog No. 5000207486 Shop All Hach Company Products
Encompass
SC1000 Probe Module-6 Sensor Connectors-8 mA/digital Input-Modbus 485-4 Relays-100-240 VAC with ConduitsIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual
Catalog No. 50-002-07486 Supplier Hach Company Supplier No. LXV400.99.1B482
SC1000 Probe Module-6 Sensor Connectors-8 mA/digital Input-Modbus 485-4 Relays-100-240 VAC with ConduitsIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual