Learn More
Hach Company SC1000 Intern Netwk Connector
Supplier:  Hach Company LZX918
Encompass
Internal network connector for the SC1000 probe module that allows for daisy chain connection of multiple probe modules Connecting cable must be purchased separately
Catalog No. 50-002-07151