Learn More
Hach Company SC1000 Mod-4 Sensor 4 mA Out
Catalog No. 5000208125 Shop All Hach Company Products
Encompass
SC1000 Probe Module-4 Sensor Connectors-4 mA Output-4 mA/digital Input-Conduit-100-240 VAC-without power cordIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual
Catalog No. 50-002-08125 Supplier Hach Company Supplier No. LXV400.99.10572
SC1000 Probe Module-4 Sensor Connectors-4 mA Output-4 mA/digital Input-Conduit-100-240 VAC-without power cordIncludesSC1000 Probe Module with installed cards (as requested)-Basic User Manual