Learn More
Hach Company SC1000 Mod-4 Sensor-4 mA Out
Catalog No. 5000207555 Shop All Hach Company Products
Encompass
SC1000 Probe Module-4 Sensor Connectors-4 mA Output-100-240 VAC-without power cordIncludessc1000 Probe Module with installed cards (as requested)-Basic User Manual
Catalog No. 50-002-07555 Supplier Hach Company Supplier No. LXV400.99.00122
SC1000 Probe Module-4 Sensor Connectors-4 mA Output-100-240 VAC-without power cordIncludessc1000 Probe Module with installed cards (as requested)-Basic User Manual