Learn More
Hach Company SC1000 Modbus License Kit
Supplier:  Hach Company LZY598
Encompass
SC1000 Modbus TCP/IP License Kit (serial nr of display module required)Serial Number (S/N) required for order The license kit LZY598 enables MODBUS TCP/IP functionality to the SC1000 display module The serial number (S/N) of the device is required to generate the license code and can be found on the back side of the SC1000 display module underneath the P/N LXG402 xx xxxxx
Catalog No. 50-002-07235