Learn More
Hach Company SC1000 Sunroof and Hardware
Supplier:  Hach Company LZX957
Encompass
SC1000 Sunroof and Mounting HardwareStainless steel pole mount kit with sunroof for SC1000 probe and display module (sc analyzer not included)
Catalog No. 50-002-07063