Learn More
TARGETMOL CHEMICALS INC SC144 25MG
SDP
Catalog No. 502576574
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SC144 is an orally active small-molecule gp130 inhibitor. purity: 98%
Catalog No. 50-257-6574 Supplier TARGETMOL CHEMICALS INC Supplier No. T620725MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SC144 is an orally active small-molecule gp130 inhibitor. purity: 98%