Learn More
Selleck Chemical LLC SC144 5mg
SDP
Supplier:  Selleck Chemical LLC S71245MG
Encompass_Preferred
SC144 is the first-in-class orally active small-moleculegp130 inhibitor that induces gp130 phosphorylation (S782) and deglycosylation, abrogates Stat3 phosphorylation and nuclear translocation, and further inhibits the expression of downstream target genes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3255