Learn More
Medchemexpress LLC HY-15614A 5mg Medchemexpress, SC144 (hydrochloride) CAS:917497-70-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15614A5MG
Encompass_Preferred
Medchemexpress, HY-15614A 5mg SC144 (hydrochloride) CAS:917497-70-2 SC144 hydrochloride is a first-in-class, orally active gp130 (IL6-beta) inhibitor. SC144 hydrochloride binds gp130, induces gp130 phosphorylation (S782) and deglycosylation, abrogates Stat3 phosphorylation and nuclear translocation, and further inhibits the expression of downstream target genes. SC144 hydrochloride shows potent inhibition of gp130 ligand-triggered signaling. SC144 hydrochloride induces apoptosis in human ovarian cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1749