Learn More
Cayman Chemical SC144 (hydrochloride), Purity: ≥98%, CAS: 917497-70-2, 10 mg
SDP
Catalog No. 502989875
Encompass_Preferred
A gp130 inhibitor; inhibits phosphorylation of STAT1, STAT3, and Akt induced by LIF and IL-6, but not IFN-γ, in OVCAR-8 ovarian cancer cells at 20 µM; induces cycle arrest at the G0/G1 phase in MDA-MB-435 breast and HT-29 colorectal cancer cells and inhibits proliferation of 14 cancer cell lines (IC50s = 0.4-4 µM); reduces tumor growth in OVCAR-8 and MDA-MB-435 mouse xenograft models
Catalog No. 50-298-9875 Supplier Cayman Chemical Supplier No. 2971410 MG
A gp130 inhibitor; inhibits phosphorylation of STAT1, STAT3, and Akt induced by LIF and IL-6, but not IFN-γ, in OVCAR-8 ovarian cancer cells at 20 µM; induces cycle arrest at the G0/G1 phase in MDA-MB-435 breast and HT-29 colorectal cancer cells and inhibits proliferation of 14 cancer cell lines (IC50s = 0.4-4 µM); reduces tumor growth in OVCAR-8 and MDA-MB-435 mouse xenograft models