Learn More
Cole-Parmer Environmental Express HotBlock SC150 Digestion System, 25-Well, 100 mL; 120 VAC
Supplier:  Cole-Parmer 5320506
Encompass
The HotBlock Systems provide faster, cleaner, and more cost-effective metals digestions, and are accepted for EPA Methods. Block heaters have an integrated controller and mat/block design and come in configurations to work directly with the HotBlock digestion cups in 15-, 50-, or 100-mL sizes.The heating blocks are made from graphite to provide excellent temperature uniformity to each well, and coated in PTFE for chemical and corrosion resistance, and to avoid sample contamination. HotBlock controllers maintain a set point to 0.2F (0.1C) and sample uniformity to 2.7F (1.5C).Compatible racks are included with each HotBlock and are designed to make sample loading and unloading as easy as possible racks have the same well design as their compatible HotBlock unit! Racks have a handle molded into each side and are made of clear polycarbonate. The digestion cups are then designed to be compatible with both the HotBlock unit and transfer racks.
Catalog No. NC1604932