Learn More
Hach Company SC1500 4 Sen 4mA Out 4 Rly
Catalog No. 5000207918 Shop All Hach Company Products
Encompass
SC1500 Controller-4 Sensor Connectors-4Ma Out-110V/Cond 4 Relay-Ext MODThe Hach SC1500 Multi-parameter Universal Controller is a state-of-the-art modular transmitter system The SC1500 enables communication for Mobile Sensor Management (MSM) solution Mobile Sensor Management provides clarity through easy access to crucial information helping you manage your process and be proactive in your maintenance This instrument connects to Claros Hach s innovative Water Intelligence System enabling you to seamlessly connect and manage instruments data and process anywhere anytime The result is greater confidence in your data and improved efficiency in your operations To unlock the full potential of Claros insist on Claros Enabled instruments
Catalog No. 50-002-07918 Supplier Hach Company Supplier No. LXV446.99.1R1M1
SC1500 Controller-4 Sensor Connectors-4Ma Out-110V/Cond 4 Relay-Ext MODThe Hach SC1500 Multi-parameter Universal Controller is a state-of-the-art modular transmitter system The SC1500 enables communication for Mobile Sensor Management (MSM) solution Mobile Sensor Management provides clarity through easy access to crucial information helping you manage your process and be proactive in your maintenance This instrument connects to Claros Hach s innovative Water Intelligence System enabling you to seamlessly connect and manage instruments data and process anywhere anytime The result is greater confidence in your data and improved efficiency in your operations To unlock the full potential of Claros insist on Claros Enabled instruments