Learn More
Hach Company SC1500 4 Sen 8xmA Out 4 Rly
Catalog No. 5000208059 Shop All Hach Company Products
Encompass
SC1500 Controller-4 Sensor Connectors-8x mA Out-110V/Cond 4 Relay-Ext MODThe Hach SC1500 Multi-parameter Universal Controller is a state-of-the-art modular transmitter system The SC1500 enables communication for Mobile Sensor Management (MSM) solution Mobile Sensor Management provides clarity through easy access to crucial information helping you manage your process and be proactive in your maintenance Claros Enabled products share measurement data and status information with Claros - The Water Intelligence System from Hach Claros provides greater confidence in your data resulting in improved efficiencies in your plant s operation To ensure you achieve the full benefits of Claros insist on Claros Enabled instruments Learn more at hach com/Claros
Catalog No. 50-002-08059 Supplier Hach Company Supplier No. LXV446.99.1R3M1
Add to Cart
Add to Cart
Request a Quote
SC1500 Controller-4 Sensor Connectors-8x mA Out-110V/Cond 4 Relay-Ext MODThe Hach SC1500 Multi-parameter Universal Controller is a state-of-the-art modular transmitter system The SC1500 enables communication for Mobile Sensor Management (MSM) solution Mobile Sensor Management provides clarity through easy access to crucial information helping you manage your process and be proactive in your maintenance Claros Enabled products share measurement data and status information with Claros - The Water Intelligence System from Hach Claros provides greater confidence in your data resulting in improved efficiencies in your plant s operation To ensure you achieve the full benefits of Claros insist on Claros Enabled instruments Learn more at hach com/Claros