Learn More
Hach Company SC200 1 analog sens HART 2 Out
Catalog No. 5000207984 Shop All Hach Company Products
Encompass
SC200 Universal Controller 100-240 V AC with one analog conductivity sensor input-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual
Catalog No. 50-002-07984 Supplier Hach Company Supplier No. LXV404.99.05202
SC200 Universal Controller 100-240 V AC with one analog conductivity sensor input-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual