Learn More
Hach Company SC200 1 anlg sens HART 2 Out
Catalog No. 5000207482 Shop All Hach Company Products
Encompass
SC200 Universal Controller 24 V DC with one analog pH/ORP/DO sensor input-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual
Catalog No. 50-002-07482 Supplier Hach Company Supplier No. LXV404.99.75102
SC200 Universal Controller 24 V DC with one analog pH/ORP/DO sensor input-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual