Learn More
Hach Company SC200 1 dig/1 anlg sens HART
Catalog No. 5000207928 Shop All Hach Company Products
Encompass
SC200 Controller-100-240 VAC-one digital sensor input-one analog pH/ORP/DO sensor input-HART-4-20 mA outputsIncludesController-Mounting Hardware-Basic User Manual
Catalog No. 50-002-07928 Supplier Hach Company Supplier No. LXV404.99.05512
SC200 Controller-100-240 VAC-one digital sensor input-one analog pH/ORP/DO sensor input-HART-4-20 mA outputsIncludesController-Mounting Hardware-Basic User Manual