Learn More
Hach Company SC200 1 dig sens HART 2 Out
Catalog No. 5000207932 Shop All Hach Company Products
Encompass
SC200 Universal Controller 24 V DC with one digital sensor input-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual
Catalog No. 50-002-07932 Supplier Hach Company Supplier No. LXV404.99.75502
SC200 Universal Controller 24 V DC with one digital sensor input-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual