Learn More
Hach Company SC200 1 dig sens In HART 2 Out
Catalog No. 5000208067 Shop All Hach Company Products
Encompass
SC200 Universal Controller 100-240 V AC with one digital sensor input-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual
Catalog No. 50-002-08067 Supplier Hach Company Supplier No. LXV404.99.05502
SC200 Universal Controller 100-240 V AC with one digital sensor input-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual