Learn More
Hach Company SC200 2 analog sens HART 2 Out
Catalog No. 5000207903 Shop All Hach Company Products
Encompass
SC200 Universal Controller 24 V DC with two analog conductivity sensor inputs-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual
Catalog No. 50-002-07903 Supplier Hach Company Supplier No. LXV404.99.75222
SC200 Universal Controller 24 V DC with two analog conductivity sensor inputs-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual