Learn More
Hach Company SC200 2 analog sens HART 2 Out
Catalog No. 5000207983 Shop All Hach Company Products
Encompass
SC200 Universal Controller 100-240 V AC with two analog pH/ORP/DO sensor inputs-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual
Catalog No. 50-002-07983 Supplier Hach Company Supplier No. LXV404.99.05112
SC200 Universal Controller 100-240 V AC with two analog pH/ORP/DO sensor inputs-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual