Learn More
Hach Company SC200 2 dig sens HART 2 Out
Catalog No. 5000208068 Shop All Hach Company Products
Encompass
SC200 Universal Controller 100-240 V AC with two digital sensor inputs-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual
Catalog No. 50-002-08068 Supplier Hach Company Supplier No. LXV404.99.05552
SC200 Universal Controller 100-240 V AC with two digital sensor inputs-HART and two 4-20mA outputsIncludesController-Mounting Hardware-Basic User Manual