Learn More
Hach Company SC200 card Modbus RS232/485
Supplier:  Hach Company 9013200
Encompass
SC200 Communication card - Modbus RS232/485The Modbus RS232/RS485 module for the SC200 is a user installed module that allows for communication via Modbus from the controller Modbus registers for Hach s probes can be downloaded from www hach com/SC200 See manual for additional connection information
Catalog No. 50-002-08101