Learn More
Hach Company SC200 Universal Controller: 100-240 V AC with one analog conductivity sensor input and two 4-20 mA outputs
Catalog No. 501997844 Shop All Hach Company Products
Encompass
The Hach SC200 Universal Controller is the most versatile controller on the market. It allows the use of both digital and analog sensors, to provide compatibility with the broadest range of sensors. The SC200 replaces the Hach SC100 digital and GLI53 analog controllers with advanced features for easier operator use. Customers may add communication modules for a variety of protocols including MODBUS RTU, Profibus DPV1, and HART. The SC200 is compatible with sensors for over 17 parameters, including pH/ORP, dissolved oxygen, chlorine, conductivity, flow, turbidity, and much more.
Catalog No. 50-199-7844 Supplier Hach Company Supplier No. LXV404.99.00202
The Hach SC200 Universal Controller is the most versatile controller on the market. It allows the use of both digital and analog sensors, to provide compatibility with the broadest range of sensors. The SC200 replaces the Hach SC100 digital and GLI53 analog controllers with advanced features for easier operator use. Customers may add communication modules for a variety of protocols including MODBUS RTU, Profibus DPV1, and HART. The SC200 is compatible with sensors for over 17 parameters, including pH/ORP, dissolved oxygen, chlorine, conductivity, flow, turbidity, and much more.