Learn More
Hach Company SC200 sun shield w/UV screen
Supplier: Hach Company 9220600
SC200 Weather and sun shield with UV protection screenThis shield is a complete solution to protect the SC200 controller from weather and sun The transparent door can be opened for operating the controller the outside case is opaque to prevent color fading of the controller case It is UV absorbent and protects the display and membrane keypad from deterioration due to UV exposure However the cover will become yellowish over time and is available as a replacement part (9221700)