Learn More
Hach Company SC4200c hole plug kit M20
Catalog No. 5000207057 Shop All Hach Company Products
Encompass
This neoprene plug can be purchased as a replacement part in order to seal any of the five open holes in the SC4200 controller The plug does not allow for cable pass through
Catalog No. 50-002-07057 Supplier Hach Company Supplier No. LXZ524.99.00031
This neoprene plug can be purchased as a replacement part in order to seal any of the five open holes in the SC4200 controller The plug does not allow for cable pass through