Learn More
Hach Company SC4200c mA out mod-5 out
Catalog No. 5000207108 Shop All Hach Company Products
Encompass
SC4200c mA output module-5 outputsIncludes When additional communications configurability is needed for process control the 4-20mA output module can be added into the controller to include five additional analog outputs This module connects to the network card on the inside of the SC4200c
Catalog No. 50-002-07108 Supplier Hach Company Supplier No. LXZ524.99.00006
SC4200c mA output module-5 outputsIncludes When additional communications configurability is needed for process control the 4-20mA output module can be added into the controller to include five additional analog outputs This module connects to the network card on the inside of the SC4200c