Learn More
Hach Company Hach SC4500 Controller (LXV525.99A11551) - Prognosys, 5x mA outputs, 2 digital sensors, 100-240 VAC
Catalog No. NC3354967 Shop All Hach Company Products
Encompass
The SC4500 is a universal digital process controller designed for water-quality monitoring systems. It interfaces with digital and analog sensors, provides configurable analog outputs and relays, and offers Ethernet and industrial protocol connectivity for local and remote monitoring and data transfer.
  • 3.5-inch TFT color display with capacitive touchpad.
  • Five configurable 0-20 mA or 4-20 mA analog outputs per analog output module.
  • Two SPDT relays with defined AC and DC switching ratings.
  • USB port for data download and software upload; records approximately 20,000 data points per sensor.
  • Panel-mount ½ DIN dimensions (144 x 144 x 192 mm) and 3.7 lb weight.
  • AC: 100-240 VAC or DC: 24 VDC power options with specified sensor load limits.
  • Metal enclosure with corrosion-resistant finish and IP66/NEMA 4X protection for panel mounting.
  • Multiple international certifications and a 12-month warranty.
Catalog No. NC3354967 Supplier Hach Company Supplier No. LXV525.99A11552
The SC4500 is a universal digital process controller designed for water-quality monitoring systems. It interfaces with digital and analog sensors, provides configurable analog outputs and relays, and offers Ethernet and industrial protocol connectivity for local and remote monitoring and data transfer.
  • 3.5-inch TFT color display with capacitive touchpad.
  • Five configurable 0-20 mA or 4-20 mA analog outputs per analog output module.
  • Two SPDT relays with defined AC and DC switching ratings.
  • USB port for data download and software upload; records approximately 20,000 data points per sensor.
  • Panel-mount ½ DIN dimensions (144 x 144 x 192 mm) and 3.7 lb weight.
  • AC: 100-240 VAC or DC: 24 VDC power options with specified sensor load limits.
  • Metal enclosure with corrosion-resistant finish and IP66/NEMA 4X protection for panel mounting.
  • Multiple international certifications and a 12-month warranty.