Learn More
Hach Company SC4500 Ctrl C1D2 Claros LAN mA
Catalog No. 5000207566 Shop All Hach Company Products
Encompass
SC4500 Controller-C1D2-Claros-enabled-LAN mA Output-1 digital Sensor-1 mA Input-100-240 VAC-without plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07566 Supplier Hach Company Supplier No. LXV525.99PAB541
SC4500 Controller-C1D2-Claros-enabled-LAN mA Output-1 digital Sensor-1 mA Input-100-240 VAC-without plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware